Learn More
BIOHIPPO Mouse VGF Microsample
SDP
Catalog No. 5000314986
Encompass
Mouse VGF Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of VGF in serum plasma tissue homogenates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 12 9 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse VGF Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse VGF Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse VGF biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14986 Supplier BIOHIPPO Supplier No. BHE15211741
Mouse VGF Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of VGF in serum plasma tissue homogenates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 12 9 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse VGF Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse VGF Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse VGF biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color