Learn More
Sino Biological Anti-Vimentin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 106411T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Mouse Vimentin/VIM, and purified by antigen affinity chromatography.
Catalog No. 50-241-1223