Learn More
Biomatik Corporation Mouse Vinculin (VCL) ELISA Kit
Supplier:  Biomatik Corporation EKU08146
Encompass_Preferred
Mouse Vinculin (VCL) ELISA Kit for detecting VCL in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.124ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q64727. Gene ID: 22330. Target Synonyms: MVCL; Metavinculin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5775