Learn More
BIOHIPPO Mouse VIP Microsample
SDP
Catalog No. 5000314290
Encompass
Mouse VIP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of VIP in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 7 82-500 pg/mL with a sensitivity of 2 93 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse VIP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse VIP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse VIP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14290 Supplier BIOHIPPO Supplier No. BHE15212233
Mouse VIP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of VIP in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 7 82-500 pg/mL with a sensitivity of 2 93 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse VIP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse VIP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse VIP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color