Learn More
Raybiotech Inc Mouse VISTA/B7-H5/PD-1H ELISA Kit
Supplier:  Raybiotech Inc ELMVISTA1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 3 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Mouse VISTA.
Catalog No. 50-195-0165