Learn More
American Research Products Inc Mouse Vitamin K dependent prot
Catalog No. 5000327164
Encompass
Mouse Vitamin K-dependent protein S (PROS1) ELISA kit from CUSABIO Detection Range 6 25 ng/mL-400 ng/mL Sensitivity 1 56 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-27164 Supplier American Research Products Inc Supplier No. CSBEL018754MO
Mouse Vitamin K-dependent protein S (PROS1) ELISA kit from CUSABIO Detection Range 6 25 ng/mL-400 ng/mL Sensitivity 1 56 ng/mL Sample serum plasma tissue homogenates Method Competitive