Learn More
Biomatik Corporation Mouse V-Ki-Ras2 Kirsten Rat Sarcoma Viral Oncogene Homolog (KRAS) ELISA Kit
Supplier:  Biomatik Corporation EKU12340
Encompass_Preferred
Mouse V-Ki-Ras2 Kirsten Rat Sarcoma Viral Oncogene Homolog (KRAS) ELISA Kit for detecting KRAS in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P32883. Gene ID: 16653. Target Synonyms: C-K-RAS; K-RAS2A; K-RAS2B; K-RAS4A; K-RAS4B; KI-RAS; KRAS1; KRAS2; NS3; RASK2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5090