Learn More
Biomatik Corporation Mouse V-Myc Myelocytomatosis Viral Oncogene Homolog (MYC) ELISA Kit
Supplier:  Biomatik Corporation EKU08179
Encompass_Preferred
Mouse V-Myc Myelocytomatosis Viral Oncogene Homolog (MYC) ELISA Kit for detecting MYC in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.066ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01108. Gene ID: 17869. Target Synonyms: C-Myc; BHLHE39; Myc Proto-Oncogene Protein; VMyc Class E basic helix-loop-helix protein 39; Transcription factor p64; Proto-oncogene c-Myc. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5189