Learn More
Biomatik Corporation Mouse Von Willebrand Factor A Domain Containing Protein 1 (vWA1) ELISA Kit
Supplier:  Biomatik Corporation EKU08189
Encompass_Preferred
Mouse Von Willebrand Factor A Domain Containing Protein 1 (vWA1) ELISA Kit for detecting vWA1 in plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8R2Z5. Gene ID: 246228. Target Synonyms: VWFA1; WARP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5185