Learn More
Sino Biological Anti-VSIG4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50187RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse VSIG4 extracellular domain (rM VSIG4; Catalog#50187-M08H; NP_808457.1; Met 1-Pro 187). VSIG4 specific IgG was purified by mouse VSIG4 affinity chromatography.
Catalog No. 50-255-3383