Learn More
Sino Biological Recombinant Mouse VSIG4 Protein (His Tag), HPLC-verified 1
Supplier:  Sino Biological 50187M08H1
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Pro 187) of mouse VSIG4 (NP_808457.1) precursor was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-224-7136