Learn More
Sino Biological Recombinant Mouse B7-H4 Protein (His Tag), Amine-Labeled Biotinylated, HPLC-verified
Catalog No. 502993633
Encompass_Preferred
A DNA sequence encoding the Mouse VTCN1 (NP_848709.2) (Phe29-Ser256) was expressed with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-299-3633 Supplier Sino Biological Supplier No. 50017M08HB100
A DNA sequence encoding the Mouse VTCN1 (NP_848709.2) (Phe29-Ser256) was expressed with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.