Learn More
Sino Biological Anti-Vitronectin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50585RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Vitronectin (Catalog#50585-M08H; NP_035837.1; Met1-Lys478). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2496