Learn More
American Research Products Inc Mouse WAP four disulfide core
Catalog No. 5000328282
Encompass
Mouse WAP four-disulfide core domain protein 2 (WFDC2) ELISA kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28282 Supplier American Research Products Inc Supplier No. CSBEL026092MO
Mouse WAP four-disulfide core domain protein 2 (WFDC2) ELISA kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates Method Sandwich