Learn More
Biomatik Corporation Mouse Wingless Type MMTV Integration Site Family, Member 3A (WNT3A) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU10978
Encompass_Preferred
Mouse Wingless Type MMTV Integration Site Family, Member 3A (WNT3A) High-sensitive ELISA Kit for detecting WNT3A in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 7.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P27467. Gene ID: 22416. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5064