Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse Wnt-3a Recombinant Protein, PeproTech®

Catalog No. 315202UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
315-20-2UG 2 μg
315-20-10UG 10 μg
31-520-100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 315-20-2UG Supplier Gibco™ Supplier No. 315202UG
  • $92.50
    $78.65 / Each of 1

    Save $13.85

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine Wnt-3a is a monomeric glycoprotein containing 334 amino acid residues. Due to glycosylation, the Murine Wnt-3a migrates at an apparent molecular weight of approximately 38.0-41.0 kDa by SDS-PAGE analysis under non-reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Wnt3a, part of the WNT gene family, consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. Study of its expression in the teratocarcinoma cell line NT2 suggests that it may be implicated in the early process of neuronal differentiation of NT2 cells induced by retinoic acid. This gene is clustered with WNT3, another family member, in the chromosome 17q21 region.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P27467
For Use With (Application) Bioactivity, Control
Formulation Protein with no preservative
Gene ID (Entrez) 22416
Molecular Weight (g/mol) 37.4 kDa
Name Mouse Wnt-3a
Purification Method Purified
Quantity 2 μg
Source Mammalian Cell
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <2 EU/μg
Gene Alias MGC119418; MGC119419; MGC119420; Protein Wnt-3a; vestigial tail; vt; wingless-related MMTV integration site 3A; wingless-type MMTV integration site family member 3A; wingless-type MMTV integration site family, member 3 like; wingless-type MMTV integration site family, member 3A; Wnt 3a; Wnt family member 3A; wnt[a]; wnt3; wnt3 l; wnt3 like; Wnt3a; Wnt-3a; WNT3A protein; wnt3l; Zwnt[a]
Common Name WNT3A
Gene Symbol WNT3A
Biological Activity Determined by its ability to induced alkaline phosphatase production by CCL-226 cells. The expected ED50 ?for this effect is 0.4-0.6 ng/ml
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence SYPIWWSLAV GPQYSSLSTQ PILCASIPGL VPKQLRFCRN YVEIMPSVAE GVKAGIQECQ HQFRGRRWNC TTVSNSLAIF GPVLDKATRE SAFVHAIASA GVAFAVTRSC AEGSAAICGC SSRLQGSPGE GWKWGGCSED IEFGGMVSRE FADARENRPD ARSAMNRHNN EAGRQAIASH MHLKCKCHGL SGSCEVKTCW WSQPDFRTIG DFLKDKYDSA SEMVVEKHRE SRGWVETLRP RYTYFKVPTE RDLVYYEASP NFCEPNPETG SFGTRDRTCN VSSHGIDGCD LLCCGRGHNA RTERRREKCH CVFHWCCYVS CQECTRVYDV HTCK
Content And Storage -20°C
Activity Determined by its ability to induced alkaline phosphatase production by CCL-226 cells. The expected ED50 ?for this effect is 0.4-0.6 ng/ml.
Expression System Mammalian cells
Form Lyophilized
Purity or Quality Grade ≥ 75% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.