Learn More
Biomatik Corporation Mouse WNT1 Inducible Signaling Pathway Protein 1 (WISP1) CLIA Kit
Supplier:  Biomatik Corporation EKU10011
Encompass_Preferred
Mouse WNT1 Inducible Signaling Pathway Protein 1 (WISP1) CLIA Kit for detecting WISP1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 2.74-2,000pg/ml. Sensitivity: 1.21pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: O54775. Gene ID: 22402. Target Synonyms: CCN4; WISP1c; WISP1i; WISP1tc; CCN family member 4; Wnt-1-induced secreted protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6936