Learn More
Biomatik Corporation Mouse X-Box Binding Protein 1 (XBP1) ELISA Kit
Supplier:  Biomatik Corporation EKU08236
Encompass_Preferred
Mouse X-Box Binding Protein 1 (XBP1) ELISA Kit for detecting XBP1 in tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O35426. Gene ID: 22433. Target Synonyms: TREB5; XBP2; Tax-responsive element-binding protein 5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8690