Learn More
Sino Biological Recombinant Anti-Lymphotactin/XCL1/SCYC1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50677R003100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse XCL1 (rM XCL1; Catalog#50677-M08B; P47993; Met1-Gly114).
Catalog No. 50-241-1682