Learn More
Sino Biological Anti-Lymphotactin/XCL1/SCYC1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50677RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse XCL1 (Catalog#50677-M08B; P47993; Met1-Gly114). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-9127