Learn More
Sino Biological Mouse XCL1 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 50677T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse XCL1 (Catalog#50677-M08B; P47993; Met1-Gly114). XCL1 specific IgG was purified by Mouse XCL1 affinity chromatography.
Catalog No. 50-237-3536