Learn More
Biomatik Corporation Mouse X-Ray Repair Cross Complementing 6 (XRCC6) CLIA Kit
Supplier:  Biomatik Corporation EKU10000
Encompass_Preferred
Mouse X-Ray Repair Cross Complementing 6 (XRCC6) CLIA Kit for detecting XRCC6 in tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 13.7-10,000pg/ml. Sensitivity: 4.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P23475. Gene ID: 14375. Target Synonyms: Ku70; CTC75; CTCBF; G22P1; ML8; TLAA; Lupus Ku Autoantigen Protein P70; ATP-Dependent DNA Helicase 2 Subunit 1; CTC box-binding factor 75 kDa; Thyroid-lupus autoantigen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8385