Learn More
Biomatik Corporation Mouse X-Ray Repair Cross Complementing 5 (XRCC5) ELISA Kit
Supplier:  Biomatik Corporation EKU08244
Encompass_Preferred
Mouse X-Ray Repair Cross Complementing 5 (XRCC5) ELISA Kit for detecting XRCC5 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P27641. Gene ID: 22596. Target Synonyms: KARP1; KU80; KUB2; Ku86; NFIV; TLAA; Lupus Ku Autoantigen Protein P80; Nuclear factor IV; 86 kDa of Ku antigen; CTC box-binding factor 85 kDa; Thyroid-lupus autoantigen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4512