Learn More
Sino Biological Anti-ZFP57 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 107454T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Mouse ZFP57, and purified by antigen affinity chromatography.
Catalog No. 50-241-1225