Learn More
Biomatik Corporation Mouse Zinc Finger Homeobox Protein 1B (ZFHX1B) CLIA Kit
Supplier:  Biomatik Corporation EKU10013
Encompass_Preferred
Mouse Zinc Finger Homeobox Protein 1B (ZFHX1B) CLIA Kit for detecting ZFHX1B in tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 13.7-10,000pg/ml. Sensitivity: 6.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9R0G7. Gene ID: 24136. Target Synonyms: ZEB2; SIP1; SIP-1; SMADIP1; SIP1; Zinc Finger E-Box Binding Homeobox 2; SMAD Interacting Protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8383