Learn More
ACROBiosystems ACROBiosystems Mouse Anti-SARS-CoV-2 Antibody IgG Titer Serologic Assay Kit (Spike Trimer)
Catalog No. 502107354
Encompass_Preferred
Kits;96TESTS;This kit is based on a standard indirect ELISA assay that allows rapid detection of Anti-SARS-CoV-2 antibodies (IgG) in mouse serum by SARS-CoV-2 Spike Protein. The kit consists of a Microplate Pre-coated with SARS-CoV-2 Spike Protein, Anti-SARS-CoV-2 Antibody (Control, Mouse IgG), and HRP-Goat-Anti-Mouse IgG Secondary Antibody.;Detection of Monoclonal Anti-SARS-CoV-2 Antibody, Mouse IgG titer by Indirect-ELISA Assay.
Immobilized SARS-CoV-2 Spike Protein at 2 μg/mL (100 μL/well) can bind Monoclonal Anti-SARS-CoV-2 Antibody, Mouse IgG in 1:100 mouse serum. Detection was performed using HRP-Anti-mouse IgG antibody with sensitivity of 20 ng/mL (QC tested).
Catalog No. 50-210-7354 Supplier ACROBiosystems Supplier No. RAST02396TESTS
Kits;96TESTS;This kit is based on a standard indirect ELISA assay that allows rapid detection of Anti-SARS-CoV-2 antibodies (IgG) in mouse serum by SARS-CoV-2 Spike Protein. The kit consists of a Microplate Pre-coated with SARS-CoV-2 Spike Protein, Anti-SARS-CoV-2 Antibody (Control, Mouse IgG), and HRP-Goat-Anti-Mouse IgG Secondary Antibody.;Detection of Monoclonal Anti-SARS-CoV-2 Antibody, Mouse IgG titer by Indirect-ELISA Assay.
Immobilized SARS-CoV-2 Spike Protein at 2 μg/mL (100 μL/well) can bind Monoclonal Anti-SARS-CoV-2 Antibody, Mouse IgG in 1:100 mouse serum. Detection was performed using HRP-Anti-mouse IgG antibody with sensitivity of 20 ng/mL (QC tested).