Learn More
World Precision Instrument WPI Standard Mouse & Rat Stereotaxic Instrument With Adaptors
SDP
Catalog No. 502539142
Encompass
For use with the WPI Rat Stereotaxic Instruments, Stoelting and most other Stereotaxic Instruments.
WPI Rat Stereotaxic Instruments with Rat and Mouse Adaptors is equipped with a U-frame base traditionally used for rat surgeries, but is supplied with rat and mouse adaptors making it the ideal stereotaxic frame to use with both species.
Available as dual, ultra-precise, digital, motorized or rotating models.
NEW FEATURE: The ONLY stereotaxic instrument line on the market with an integrated warming base included on all units! Simply purchase the Temperature Control Box 1 (505375), or Temperature Control Box 2 (505376) and probe (505377) to use the warming function.
View our Stereotaxic Frame Compatibility Chart
Catalog No. 50-253-9142 Supplier World Precision Instrument Supplier No. 505281
Add to Cart
Add to Cart
Request a Quote
For use with the WPI Rat Stereotaxic Instruments, Stoelting and most other Stereotaxic Instruments.
WPI Rat Stereotaxic Instruments with Rat and Mouse Adaptors is equipped with a U-frame base traditionally used for rat surgeries, but is supplied with rat and mouse adaptors making it the ideal stereotaxic frame to use with both species.
Available as dual, ultra-precise, digital, motorized or rotating models.
NEW FEATURE: The ONLY stereotaxic instrument line on the market with an integrated warming base included on all units! Simply purchase the Temperature Control Box 1 (505375), or Temperature Control Box 2 (505376) and probe (505377) to use the warming function.
View our Stereotaxic Frame Compatibility Chart