Learn More
World Precision Instrument WPI New Standard Mouse & Rat Stereotaxic, Warm Base Only
SDP
Catalog No. 502539206
Encompass
Like the WPI Rat Stereotaxic Instruments, the WPI Mouse and Rat Stereotaxic Instrument line has larger laser engraved vernier scales (accurate to 100 um), 80 mm of ventral-dorsal, medial-lateral and anterior-posterior travel, absolute lock at 90° ventral-dorsal and brass-bushings for electrical grounding. It is available as a digital or motorized model.
View our Stereotaxic Frame Compatibility Chart
Catalog No. 50-253-9206 Supplier World Precision Instrument Supplier No. 505205
Like the WPI Rat Stereotaxic Instruments, the WPI Mouse and Rat Stereotaxic Instrument line has larger laser engraved vernier scales (accurate to 100 um), 80 mm of ventral-dorsal, medial-lateral and anterior-posterior travel, absolute lock at 90° ventral-dorsal and brass-bushings for electrical grounding. It is available as a digital or motorized model.
View our Stereotaxic Frame Compatibility Chart