Learn More
Cayman Chemical MousentIDYKDDDDKIgGDyLIg1ea
Supplier: Cayman Chemical 112561 EA
Mouse anti-DYKDDDDK IgG (Clone M2) conjugated to DyLight 550 Antibody: Mouse monoclonal anti-DYDDDDK IgG Clone: M2 Isotype: IgG Dye: DyLight 550 Excitation max. λ: 562 nm Emission max. λ: 576 nm Uses: Fluorescence-activated cell sorting, flow cytometry, microscopy, and protein microarrays