Learn More
Cayman Chemical MousentIDYKDDDDKIgGDyLIg1ea
SDP
Catalog No. 502951100
Encompass_Preferred
Mouse anti-DYKDDDDK IgG (Clone M2) conjugated to DyLight 488 Antibody: Mouse monoclonal anti-DYDDDDK IgG Clone: M2 Isotype: IgG Dye: DyLight 488 Excitation max. λ: 493 nm Emission max. λ: 518 nm Uses: Fluorescence-activated cell sorting, flow cytometry, and protein microarrays
Catalog No. 50-295-1100 Supplier Cayman Chemical Supplier No. 112481 EA
Mouse anti-DYKDDDDK IgG (Clone M2) conjugated to DyLight 488 Antibody: Mouse monoclonal anti-DYDDDDK IgG Clone: M2 Isotype: IgG Dye: DyLight 488 Excitation max. λ: 493 nm Emission max. λ: 518 nm Uses: Fluorescence-activated cell sorting, flow cytometry, and protein microarrays