Learn More
Cayman Chemical MousentIDYKDDDDKIgGSureL1mg
SDP
Catalog No. 502953817
Encompass_Preferred
Mouse anti-DYKDDDDK IgG (Clone M2) conjugated to SureLight APC Antibody: Mouse monoclonal anti-DYKDDDDK IgG Clone: M2 Dye: SureLight™ Allophycocyanin (APC) Excitation max. λ: 652 nm Emission max. λ: 657.5 nm Uses: Flow cytometry/Fluorescence-activated cell sorting, High-throughput screening, Time-resolved fluorescence energy transfer
Catalog No. 50-295-3817 Supplier Cayman Chemical Supplier No. 134041 MG
Mouse anti-DYKDDDDK IgG (Clone M2) conjugated to SureLight APC Antibody: Mouse monoclonal anti-DYKDDDDK IgG Clone: M2 Dye: SureLight™ Allophycocyanin (APC) Excitation max. λ: 652 nm Emission max. λ: 657.5 nm Uses: Flow cytometry/Fluorescence-activated cell sorting, High-throughput screening, Time-resolved fluorescence energy transfer