Learn More
Braintree Scientific MouseOx Plus Multiplexer - 8 Channel w/temperature
SDP
Catalog No. 501955921
Encompass
MouseOx Plus Add-on, Multiplexer Sensor Switching Unit for MouseOx Plus. Allows 1 MouseOx Plus to monitor and record data for up to 16 subjects. The MouseOx Plus Multiplexer is an accessory that allows you to monitor multiple subjects, one at a time, using one MouseOx Plus control unit . The Multiplexer allows you to choose between automatic or manual switching. Summary screen shows all subjects being monitored by the Multiplexer as well as all physiological parameters reported by the MouseOx Plus **Sold for research purposes only and not for human use**
Catalog No. 50-195-5921 Supplier Braintree Scientific Supplier No. SLS015040
MouseOx Plus Add-on, Multiplexer Sensor Switching Unit for MouseOx Plus. Allows 1 MouseOx Plus to monitor and record data for up to 16 subjects. The MouseOx Plus Multiplexer is an accessory that allows you to monitor multiple subjects, one at a time, using one MouseOx Plus control unit . The Multiplexer allows you to choose between automatic or manual switching. Summary screen shows all subjects being monitored by the Multiplexer as well as all physiological parameters reported by the MouseOx Plus **Sold for research purposes only and not for human use**