Learn More
Abcam Mouse/Rat ACTH ELISA Kit (adrenocorticotropic hormone), 1 x 96 Tests
Catalog No. 502656716
Encompass_Preferred
Mouse/Rat ACTH ELISA Kit (adrenocorticotropic hormone) is a single-wash 90-min Simplestep used to quantify Mouse/Rat ACTH (adrenocorticotropic hormone) with a sensitivity of 6 pg/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10x96 plates for bulk orders - Design your own immunoassay: we also offer the conjugation-ready antibody pair - Cited in over 10 citations
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-6716 Supplier Abcam Supplier No. AB2638801X96TESTS
Mouse/Rat ACTH ELISA Kit (adrenocorticotropic hormone) is a single-wash 90-min Simplestep used to quantify Mouse/Rat ACTH (adrenocorticotropic hormone) with a sensitivity of 6 pg/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10x96 plates for bulk orders - Design your own immunoassay: we also offer the conjugation-ready antibody pair - Cited in over 10 citations
The product is subject to the following: Abcam Restricted Use Statement