Learn More
Abcam Mouse/Rat Cathepsin B Antibody Pair - BSA and Azide free, 10 x 96 Tests
Catalog No. 502654345
Encompass_Preferred
Mouse/Rat Cathepsin B Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Mouse/Rat Cathepsin B.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-4345 Supplier Abcam Supplier No. AB30017510X96TESTS
Mouse/Rat Cathepsin B Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Mouse/Rat Cathepsin B.
The product is subject to the following: Abcam Restricted Use Statement