Learn More
Kent Scientific MouseSTAT™ Jr. Stand-Alone Pulse Oximeter
SDP
Catalog No. 13005101 Shop All Kent Scientific Products
Encompass
Change view
Click to view available options
Includes:
controller and power supply
Catalog No. Includes
13-005-101 controller and power supply
1 options
Catalog No. 13-005-101 Supplier Kent Scientific Supplier No. MSTATJR
Includes
Controller (sensors sold separately)
Helps provide accurate heart rate and pulse oximetry measurements in both mice and rats.
  • For measurement in mice and rats
  • Two displays for heart rate and pulse oximetry
  • Measures heart rates up to 900bpm
  • Visual alerts to confirm sensor placement
Specifications
For Use With (Application) Heart rate and pulse oximetry measurements in mice and rats
Type Pulse Oximeter
Includes controller and power supply