Learn More
Kent Scientific MouseSTAT™ Pulse Oximeter Paw Sensor
SDP
Catalog No. 13005134 Shop All Kent Scientific Products
Encompass
Change view
Click to view available options
Type:
Pulse Oximeter Paw Sensor
Pulse Oximeter Paw Sensor/MRI Comp.
Animal:
Rat, Pup
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Animal
13-005-134 Pulse Oximeter Paw Sensor/MRI Comp. Rat, Pup
13-005-133 Pulse Oximeter Paw Sensor Rat, Pup
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-005-134 Supplier Kent Scientific Supplier No. SPO25MMMRI
For use with PhysioSuite and PhysioSuite Modules.
  • Recommended for use with rats and pups
  • MRI compatible sensor includes 3m cable
Applications
Vital sign monitoring, pulse oximetry, heart rate, physiological monitoring
Specifications
Animal Rat, Pup
For Use With (Application) Vital Sign Monitoring, Pulse Oximetry, Heart Rate, Physiological Monitoring
Type Pulse Oximeter Paw Sensor/MRI Comp.
For Use With (Equipment) PhysioSuite and PhysioSuite Modules
Includes 3m Cable