Learn More
Cayman Chemical Mouse/Rat Peroxisome Proliferator-Activated Receptor Gamma Reporter Assay System, 3x32 Well
SDP
Catalog No. 502972570
Encompass_Preferred
mrPPARγ Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields exceptional cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for cumbersome intermediate treatment steps such as spin-and-rinse of cells, viability determinations, or cell titer adjustments prior to assay setup. [INDIGO Catalog Nos. MR00101-32, MR00101]
Catalog No. 50-297-2570 Supplier Cayman Chemical Supplier No. 157613X32 WELL
mrPPARγ Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields exceptional cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for cumbersome intermediate treatment steps such as spin-and-rinse of cells, viability determinations, or cell titer adjustments prior to assay setup. [INDIGO Catalog Nos. MR00101-32, MR00101]