Learn More
Alzet ALZETTM Mouse Jugular Catheter: Optimized for Jugular Delivery from any ALZET Osmotic Pump
SDP
Catalog No. NC9382176
Encompass
This ALZET catheter connects to any ALZET Osmotic Pump to enable continuous IV delivery of test agents in mice. EtO sterilized and ready for immediate use.Catheter Material: PolyurethaneSpecifications: 9 cm total length; distal tip 1 Fr; internal volume ~10 μlOptimum Animal Size: 30+ grams (insertion marks for adaptation to smaller animals)Features & Benefits:•Retention beads at 1.3 and 3.8 cm from the tip facilitate anchoring to tissue•ALZET connection provides secure fit to all pump models•Catheter tip optimum for use with other small vessels (e.g., femoral, carotid, etc.)•Marked at 0.9 and 1.1 cm to simplify adaptation for animals weighing <25 g and 25-30 g respectively
Catalog No. NC9382176 Supplier Alzet Supplier No. 0007700
This ALZET catheter connects to any ALZET Osmotic Pump to enable continuous IV delivery of test agents in mice. EtO sterilized and ready for immediate use.Catheter Material: PolyurethaneSpecifications: 9 cm total length; distal tip 1 Fr; internal volume ~10 μlOptimum Animal Size: 30+ grams (insertion marks for adaptation to smaller animals)Features & Benefits:•Retention beads at 1.3 and 3.8 cm from the tip facilitate anchoring to tissue•ALZET connection provides secure fit to all pump models•Catheter tip optimum for use with other small vessels (e.g., femoral, carotid, etc.)•Marked at 0.9 and 1.1 cm to simplify adaptation for animals weighing <25 g and 25-30 g respectively