Learn More
SAI Infusion Technologies Rodent Tail Vein Catheter - 28cm, 0.75 F tip
SDP
Catalog No. NC0250640
Encompass
28cm PU Tail Vein Catheter with a 0.75F (32G) tip, and female luer for easy syringe access. Sterile. This SAI tail vein access catheter easily inserts into the tail vein of a restrained rodent without surgery. The size allows for quick, mistake free access to the tail vein, and once inserted, the flexibility keeps vessel trauma to a minimum. Unlike sharp, stiff needles this catheter moves freely up the vein to ensure your dose goes into the bloodstream, not into the surrounding tissue or back out through the insertion site. It bends naturally with the tail, reducing injury and allowing for continuous infusion or repeated bolus dosing with less chance of losing the tail. Great for short term access in restrained or anesthetized subjects. Product is for laboratory animal research only.
Catalog No. NC0250640 Supplier SAI Infusion Technologies Supplier No. MTV11
28cm PU Tail Vein Catheter with a 0.75F (32G) tip, and female luer for easy syringe access. Sterile. This SAI tail vein access catheter easily inserts into the tail vein of a restrained rodent without surgery. The size allows for quick, mistake free access to the tail vein, and once inserted, the flexibility keeps vessel trauma to a minimum. Unlike sharp, stiff needles this catheter moves freely up the vein to ensure your dose goes into the bloodstream, not into the surrounding tissue or back out through the insertion site. It bends naturally with the tail, reducing injury and allowing for continuous infusion or repeated bolus dosing with less chance of losing the tail. Great for short term access in restrained or anesthetized subjects. Product is for laboratory animal research only.