Learn More
Fritsch Milling And Sizing Inc Movable Jaw P-1 Pl+Cl Size 2, Mn-Steel
SDP
Catalog No. 501534397
Encompass
Movable crushing jaw, manganese steel for PULVERISETTE 1 premium line and classic line Size 2 for the comminution of hard, brittle samples
General material specifications:
Material: manganese steel - 1.3401
Abrasion resistance: good
Suitable for: hard, brittle samples
Catalog No. 50-153-4397 Supplier Fritsch Milling And Sizing Inc Supplier No. 43.3140.23
Movable crushing jaw, manganese steel for PULVERISETTE 1 premium line and classic line Size 2 for the comminution of hard, brittle samples
General material specifications:
Material: manganese steel - 1.3401
Abrasion resistance: good
Suitable for: hard, brittle samples