Learn More
Fritsch Milling And Sizing Inc Movable Jaw P-1 Cl Size 1 Chromium-Free Tool Steel
SDP
Catalog No. 501534367
Encompass
Movable crushing jaw, chromium-free tool steel for PULVERISETTE 1 classic line, Size 1 for the comminution of medium-hard samples
General material specification:
Material: chromium-free steel
Abrasion resistance: good
Suitable for: medium-hard samples
Catalog No. 50-153-4367 Supplier Fritsch Milling And Sizing Inc Supplier No. 43.0021.09
Movable crushing jaw, chromium-free tool steel for PULVERISETTE 1 classic line, Size 1 for the comminution of medium-hard samples
General material specification:
Material: chromium-free steel
Abrasion resistance: good
Suitable for: medium-hard samples