Learn More
Fritsch Milling And Sizing Inc Movable Jaw P-1 Cl Size 1 Hardmetal Tungsten Carbide
SDP
Catalog No. 501534371
Encompass
Movable crushing jaw, hardmetal tungsten carbide for PULVERISETTE 1 classic line, Size 1 for the comminution of hard, abrasive samples
General material specifications:
Material: hardmetal tungsten carbide - GC40 - WC
Abrasion resistance: extremely good
Suitable for: hard, abrasive samples
Catalog No. 50-153-4371 Supplier Fritsch Milling And Sizing Inc Supplier No. 43.0060.08
Add to Cart
Add to Cart
Request a Quote
Movable crushing jaw, hardmetal tungsten carbide for PULVERISETTE 1 classic line, Size 1 for the comminution of hard, abrasive samples
General material specifications:
Material: hardmetal tungsten carbide - GC40 - WC
Abrasion resistance: extremely good
Suitable for: hard, abrasive samples