Learn More
Fritsch Milling And Sizing Inc Movable Jaw P-1 Cl Size 1 Manganese Steel
SDP
Catalog No. 501534379
Encompass
Movable crushing jaw, manganese steel for PULVERISETTE 1 classic line, Size 1 for the comminution of hard, brittle samples
General material specification:
Material: manganese steel - 1.3401
Abrasion resistance: good
Suitable for: hard, brittle samples
Catalog No. 50-153-4379 Supplier Fritsch Milling And Sizing Inc Supplier No. 43.0140.23
Movable crushing jaw, manganese steel for PULVERISETTE 1 classic line, Size 1 for the comminution of hard, brittle samples
General material specification:
Material: manganese steel - 1.3401
Abrasion resistance: good
Suitable for: hard, brittle samples