Learn More
Fritsch Milling And Sizing Inc Movable Jaw P-1 Pl+Cl Size 2 Chromium-Free Tool Steel
SDP
Catalog No. 501534385
Encompass
Movable crushing jaw, chronium-free tool steel for PULVERISETTE 1 premium line and classic line, Size 2 for the comminution of medium-hard samples
General material specifications:
Material: chromium free steel
Abrasion resistance: good
Suitable for: medium-hard samples
Catalog No. 50-153-4385 Supplier Fritsch Milling And Sizing Inc Supplier No. 43.3021.09
Movable crushing jaw, chronium-free tool steel for PULVERISETTE 1 premium line and classic line, Size 2 for the comminution of medium-hard samples
General material specifications:
Material: chromium free steel
Abrasion resistance: good
Suitable for: medium-hard samples