Learn More
Fritsch Milling And Sizing Inc Movable Jaw P-1 Pl+Cl Size 2 Stainless Steel
SDP
Catalog No. 501534387
Encompass
Movable crushing jaw, stainless steel for PULVERISETTE 1 premium line and classic line, Size 2 for the comminution of medium-hard, brittle samples
General material specification:
Material: stainless steel - 1.4571
Abrasion resistance: fairly good
Suitable for: medium-hard, brittle samples
Catalog No. 50-153-4387 Supplier Fritsch Milling And Sizing Inc Supplier No. 43.3040.10
Movable crushing jaw, stainless steel for PULVERISETTE 1 premium line and classic line, Size 2 for the comminution of medium-hard, brittle samples
General material specification:
Material: stainless steel - 1.4571
Abrasion resistance: fairly good
Suitable for: medium-hard, brittle samples