Learn More
Fritsch Milling And Sizing Inc Movable Jaw P-1 Pl+Cl Size 2 Hardmetal Tungsten Carbide
SDP
Catalog No. 501534389
Encompass
Movable crushing jaw, hardmetal tungsten carbide for PULVERISETTE 1 premium line and classic line, Size 2
For the comminution of hard, abrasive samples
General material specifications:
Material: hardmetal tungsten carbide - GC 40 - WC
Abrasion resistance: very good
Suitable for hard, abrasive samples
Catalog No. 50-153-4389 Supplier Fritsch Milling And Sizing Inc Supplier No. 43.3060.08
Add to Cart
Add to Cart
Request a Quote
Movable crushing jaw, hardmetal tungsten carbide for PULVERISETTE 1 premium line and classic line, Size 2
For the comminution of hard, abrasive samples
General material specifications:
Material: hardmetal tungsten carbide - GC 40 - WC
Abrasion resistance: very good
Suitable for hard, abrasive samples