Learn More
Fritsch Milling And Sizing Inc Movable Jaw P-1 Pl+Cl Size 2 Hardened Steel
SDP
Catalog No. 501534384
Encompass
Movable crushing jaw, hardened steel for PULVERISETTE 1 premium line and classic line, Size 2 for the comminution of brittle, very hard samples
Crushing jaws made of hardened steel belong to the standard equipment of the FRITSCH Jaw Crushers and therefore they are included in the instrument price.
General material specification:
Material: hardened steel - 1.2080
Abrasion resistance: good
suitable for: brittle, very hard samples
Catalog No. 50-153-4384 Supplier Fritsch Milling And Sizing Inc Supplier No. 43.3020.09
Movable crushing jaw, hardened steel for PULVERISETTE 1 premium line and classic line, Size 2 for the comminution of brittle, very hard samples
Crushing jaws made of hardened steel belong to the standard equipment of the FRITSCH Jaw Crushers and therefore they are included in the instrument price.
General material specification:
Material: hardened steel - 1.2080
Abrasion resistance: good
suitable for: brittle, very hard samples