Learn More
ATCC MOVAS; Aortic Smooth Muscle; Mouse (Mus musculus)
Catalog No. 502383184
Encompass
MOVAS is a spindle-shaped cell line that was isolated in 1999 from the smooth muscle of an adult mouse. This cell line was deposited by M Husain and can be used in cardiovascular disease research.
Catalog No. 50-238-3184 Supplier ATCC Supplier No. CRL2797
Add to Cart
Add to Cart
MOVAS is a spindle-shaped cell line that was isolated in 1999 from the smooth muscle of an adult mouse. This cell line was deposited by M Husain and can be used in cardiovascular disease research.