Learn More
TARGETMOL CHEMICALS INC MOVELTIPRIL 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T258325MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Moveltipril (Moveltipril calcium) is a potent angiotensin-converting enzyme (ACE) inhibitor.Moveltipril is converted to captopril in the body for action. purity: 99%
Catalog No. 50-258-0737