Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Mowiol™ 4-88

Catalog No. 8138150G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
81-381-50G 50 g
81-381-1KG 1 kg
81-381-250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 81-381-50G Supplier Sigma Aldrich Supplier No. 8138150G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Mowiol(R) is a water soluble hydrocolloid mucoadhesive based on poly (vinyl alcohol). Mowiol(R) at pH 8.5 has a couple of advantages over other mounting media:its a liquid when the sample is mounted and solidifies after several hours, forming a permanent bond between the coverslip and slidefluorescence is stable when held in dark and at 4oC.Mowiol(R) is used as an embedding medium for preserving stained cell dishes in the study of siRNA array production of HeLa cells. Mowiol(R) is also reported to be used as a mounting medium in:design of TSAR and its applicationsdetection of the secreted and cytoplasmic fractions of IpaB, IpaC and IpaD by lysozyme permeabilizationfluorescence recovery after photobleaching (FRAP).Mowiol(R) may be used as a coating for sensors based on piezoelectric quartz crystals. Another probable use of Mowiol(R) is during the study of super fluorescence imaging of biocompatible carbon dots.Mowiol is a registered trademark of Kuraray Europe GmbH

Specifications

CAS 9002-89-5
Linear Formula [-CH2CHOH-]n
MDL Number MFCD00081922
Quantity 50 g
Synonym Poly(vinyl alcohol)
Molecular Weight (g/mol) 31000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.